Please login to your online account to display your discounted pricing

Troemner™ Individual Analytical Precision Weights, Class 1 with Traceable Certificate


Designed for your most demanding weighing applications

Manufacturer: troemner llc  70171T

 View more versions of this product

Catalog No. 02-319-1AT

  • / Each
 

Description & Specifications

Specifications

Product Type Metric analytical weight with handling knob
Certifications/Compliance Traceable certificate, ISO9001
Includes Polycarbonate case
Classification ASTM Class 1
Quantity Each
Packaging Quantity 1
Material 316 Stainless Steel
Weight (Metric) 100g
Density 7.95g/cm3 at 20°C
Mass 100g
Shape Cylindrical with handling knob