Please login to your online account to display your discounted pricing

Troemner™ Individual Analytical Precision Weights, Class 1 with Traceable Certificate


Designed for your most demanding weighing applications

Manufacturer: fisher scientific  70261T

 View more versions of this product

Catalog No. 02-319-1HT

  • / Each
 

Description & Specifications

Specifications

Certifications/Compliance Traceable certificate, ISO9001
Includes Polycarbonate case
Classification ASTM Class 1
Quantity Each
Packaging Quantity 1
Weight (Metric) 500mg
Density 7.95g/cm3 at 20°C
Mass 500mg
Shape Flat with one side turned up
Product Type Metric analytical sheet weight with one side turned up
Material 304 Stainless Steel