Please login to your online account to display your discounted pricing

Troemner™ Individual Analytical Precision Weights, Class 1 with Statement of Accuracy


Low magnetic susceptibility, uniform consistency and high resistance to corrosion

Manufacturer: fisher scientific  70321F

 View more versions of this product

Catalog No. 02-319-1N

  • / Each
 

Description & Specifications

Specifications

Product Type One-piece metric analytical sheet weight
Certifications/Compliance Statement of accuracy, ISO9001
Includes Polycarbonate case
Classification ASTM Class 1
Quantity Each
Packaging Quantity 1
Material 304 Stainless Steel
Weight (Metric) 20mg
Density 7.95g/cm3 at 20°C
Mass 20mg
Shape Flat with one end turned up for easy handling
Accessories

Accessories for Catalog Number: 023191N

Item Mfr. No. Item Description Price Per Unit Quantity