Please login to your online account to display your discounted pricing

Troemner™ Individual Analytical Precision Weights, Class 1 with NVLAP Certificate


Low magnetic susceptibility, uniform consistency and high resistance to corrosion

Manufacturer: fisher scientific  70261W

 View more versions of this product

Catalog No. 02-319-1HC

  • / Each
 

Description & Specifications

Specifications

Quantity Each
Classification ASTM Class 1
Shape Flat with one end turned up for easy handling
Material 304 Stainless Steel
Density 7.95g/cm3 at 20°C
Certifications/Compliance NVLAP, ISO9001
Includes Polycarbonate case
Packaging Quantity 1
Weight (Metric) 500mg
Mass 500mg
Product Type One-piece metric analytical sheet weight