Troemner™ Heavy Duty Dual Adjust Small Tapered Clamps
Designed to hold vessels with ground glass joint necks
$360.06 - $360.06
Specifications
Material Nickel plated zinc
Product Type Tapered Clamp
View More Specs
Products 2
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
02-216-202 View Documents Troemner™
915290
0.6mm (0.02 in.) Each for $360.06
02-216-203 View Documents Troemner™
915291
0.756mm (0.03 in.) Each for $360.06
Description
Description
  • Clamps are constructed with extension rods for easy attachment to lab-frames and other apparatus
  • Nickel-plated zinc construction
Specifications
Specifications
Nickel plated zinc
Tapered Clamp