missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Troemner Electric Balance Weight Cases

Designed to securely hold and protect weights sized 2kg through 10g

Manufacturer:  Troemner Llc SWCE0500

 View more versions of this product

Catalog No. 02-215-540


 

Specifications Description & Specifications

Specifications

8.9cm
Securely holds, protects and transports weights
500g weight
Handle, a recessed grip, side latches and a color-coded insert
4.5 in.
11.4cm
Individual electronic balance weight case
3.5 in.
Each
Polycarbonate