Troemner™ Troemner Electric Balance Weight Cases
Designed to securely hold and protect weights sized 2kg through 10g
Manufacturer:  Troemner™ SWCE0160
 View more versions of this product
Catalog No. 02-215-535
Description
Description
  • Lid is rotated open, color-coded insert and weight move with lid
  • Prevents the urethane bumper from rubbing on weight, further protecting weight's calibration
Specifications
Specifications
Individual electronic balance weight case
2 in.
2.5 in.
Securely holds, protects and transports weights
Handle, a recessed grip, side latches and a color-coded insert
Polycarbonate
5.1cm
6.4cm
160g weight
Each
Documents