missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Troemner™ Electric Balance Weight Cases

Designed to securely hold and protect weights sized 2kg through 10g

Supplier:  Troemner™ 80864243

 View more versions of this product

Catalog No. 02-215-541



Description

Description

  • Lid is rotated open, color-coded insert and weight move with lid
  • Prevents the urethane bumper from rubbing on weight, further protecting weight's calibration

Specifications

Specifications

Individual Electronic Balance Weight Case
3.5 in.
4.5 in.
Securely holds, protects and transports weights
Handle, a recessed grip, side latches and a color-coded insert
Polycarbonate
8.9 cm
11.4 cm
600g weight
Each
SDS
Documents

Documents

Promotions

Promotions