Troemner™ Cal-Paks™ Weight Sets: Class 1
Highly polished Alloy 8 stainless-steel weights
Manufacturer:  Troemner™ 72681W
 View more versions of this product
Catalog No. 02-215-622
Includes: Master carrying case; three individually cased precision weights; USB Flashdrive with electronic NVLAP™ certification; NVLAP Lab Code 105013-0; NVLAP certificate; and accessories for proper handling and weight care
Description
Description
Highly polished Alloy 8 stainless-steel weights
Compliance
Meet ISO™/IEC 17025 requirements.
Specifications
Specifications
Large Weight Set
ASTM Class 1
7.95g/cm3 at 20°C
Cylindrical weight: 5kg, 200g, 2g, Polycarbonate case in Polypropylene case; NVLAP certificate
3
NVLAP, ISO9001
5202g
2g, 200g, 5kg
3/Set
Documents