Troemner™ Cal-Paks™ Weight Sets: Class 1
Highly polished Alloy 8 stainless-steel weights
Manufacturer:  Troemner™ 72711W
 View more versions of this product
Catalog No. 02-215-619
Includes: Master carrying case; three individually cased precision weights; USB Flashdrive with electronic NVLAP™ certification; NVLAP Lab Code 105013-0; NVLAP certificate; and accessories for proper handling and weight care
Description
Description
Highly polished Alloy 8 stainless-steel weights
ANSI™/ASTM™ Class 1.
  • Components and case are linked through the use of a unique serial number
  • Clear plastic case
  • Individual weights in high-quality polycarbonate cases
Compliance
Meet ISO™/IEC 17025 requirements.
Specifications
Specifications
Small Weight Set
ASTM Class 1
7.95g/cm3 at 20°C
Cylindrical weight: 20g, 2g, sheet weight: 200mg, Polycarbonate case; NVLAP certificate
3
NVLAP, ISO9001
22.2g
200mg, 2g and 20g
3/Set
Documents