Troemner™ Cal-Paks™ Weight Sets: Class 1
Highly polished Alloy 8 stainless-steel weights
Manufacturer:  Fisher Scientific 72561W
 View more versions of this product
Catalog No. 02-215-606
Specifications Description & Specifications
Specifications
Cal-Paks™ small weight set
ASTM Class 1
7.95g/cm3 at 20°C
Cylindrical weight: 100g, 10g, 2g, Polycarbonate case; NVLAP certificate
3
NVLAP, ISO9001
112g
2g, 10g, 100g
3/Set