Please login to your online account to display your discounted pricing
Troemner™ Cal-Paks™ Weight Sets: Class 0
Highly polished Alloy 8 stainless-steel weights
$687.71 - $741.74
Specifications
Certifications/Compliance NVLAP, ISO9001
Quantity 3/Set
View More Specs
Includes:  Master carrying case; three individually cased precision weights; USB Flashdrive with electronic NVLAP™ certification; NVLAP Lab Code 105013-0; NVLAP certificate; and accessories for proper handling and weight care
Products
Catalog Number Mfr. No. Includes Mass Price Quantity    
02-215-600
troemner llc
725000W
Troemner Alloy 8 cylindrical weight: 2g, Stainless Steel weight: 200mg, Aluminuml weight: 1mg, Polycarbonate case; NVLAP certificate 1mg, 200mg, 2g Each for $687.71
02-215-601
troemner llc
725100W
Troemner Alloy 8. cylindrical weight: 5g, Stainless Steel weight: 500mg, Aluminuml weight: 1mg, Polycarbonate case; NVLAP certificate 1mg, 500mg, 5g Each for $741.74
Description & Specifications
Specifications
Certifications/Compliance NVLAP, ISO9001
Quantity 3/Set
ANSI™/ASTM™ Class 0.
  • Components and case are linked through the use of a unique serial number
  • Clear plastic case
  • Individual weights in high-quality polycarbonate cases
Meet ISO™/IEC 17025 requirements.