missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Troemner™ Analytical Weight Cases

Polycarbonate cases securely hold and protect ASTM weights of various sizes

Manufacturer:  Troemner™ SWCA0010

 View more versions of this product

Catalog No. 02-215-551


 

Description

Description

  • Designed to hold and protect weights sized 2kg through 10g
  • Lid is rotated open, color-coded insert and weight move with lid
  • Prevents urethane bumper from rubbing on weight, further protecting weight's calibration

Specifications

Specifications

Individual analytical weight case
2 in.
10g weight
Polycarbonate
2.5 in.
Each
Documents