Please login to your online account to display your discounted pricing

Troemner™ Analytical Weight Cases


Polycarbonate cases securely hold and protect ASTM weights of various sizes

Manufacturer: troemner llc  SWCA2000

 View more versions of this product

Catalog No. 02-215-560

  • / Each
 

Description & Specifications

Specifications

Quantity Each
Height (English) Case 4.5 in.
For Use With (Equipment) 2kg weight
Diameter (English) Outside 3.5 in.
Product Type Individual analytical weight case
Material Polycarbonate