missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Troemner™ Alloy 8 Metric Precision Weight Sets, UltraClass

Excellent for high-precision calibrations and reference standards

Manufacturer:  Troemner™ 72240F

 View more versions of this product

Catalog No. 02-214-3A



Includes: Forceps

Description

Description

Excellent for high-precision calibrations and reference standards

Manufactured from a specially developed austenitic stainless-steel—Alloy 8—available exclusively from Troemner. Weights 1g and larger are manufactured using Alloy 8, which possesses closely controlled density, extremely low magnetic susceptibility, excellent stability and resistance to corrosion, assuring a very stable mass over time. Other weights are stainless steel.

  • Each fractional weight is embossed with its value
  • Weights from 1mg to 30mg are aluminum and flat, with one side turned up to form a handle for non-slip gripping with forceps
  • Weights from 50 to 500mg are stainless steel and flat with one side turned up to form a handle
  • Gram and kilogram weights are manufactured using Alloy 8 and come in cylindrical shapes with knobs on top for easy handling
  • Weight sets come in a 5-2-2-1 series, with two weights of any included mg or g value beginning with “2”quotation (2mg, 20mg, etc.) and one weight for all other inclusive values
  • Each weight set (100g to 1mg) is packaged in a polypropylene case
  • Weight sets 500g and above come packaged in a wood case
  • All weights come with a statement of accuracy

Certifications

A NIST/NVLAP™ weight calibration certificate or traceable certificate may be purchased to meet all of your ISO™, GLP™, GMP™, FDA, or nuclear requirements.

Specifications

Specifications

Medium metric analytical weight set
Stainless Steel
1g to 100g
9/Set
UltraClass
1g to 100g
Cylindrical weight: 100g, 50g, 2 x 20g, 10g, 5g, 2 x 2g, 1g, Polycarbonate case
Multiple
Documents
Accessories