Troemner™ Alloy™ 8 Metric Precision Weight Sets, UltraClass with Statement of Accuracy
Excellent for high-precision calibrations and reference standards
Manufacturer:  Troemner Llc 72210
 View more versions of this product
Catalog No. 02-215-119
Specifications Description & Specifications
Specifications
1mg to 200g
UltraClass
Multiple
Alloy 8 Stainless Steel, Aluminum
1mg to 200g
Stainless Steel cylindrical weight: 2 x 200g, 100g, 50g, 2 x 20g, 10g, 5g, 2 x 2g, 1g, Stainless Steel sheet weight: 500mg, 2 x 200mg, 100mg, 50mg, 2 x 20mg, 10mg, 5mg, Aluminium sheet weight: 2 x 2mg, 1mg, Polycarbonate case
23/Set
Medium metric analytical weight set
Statement of accuracy, ISO9001
Accessories