Please login to your online account to display your discounted pricing
Troemner™ Alloy 8 Metric Precision Weight Sets, Class 4 with Statement of Accuracy
Low magnetic susceptibility, uniform consistency and high resistance to corrosion
Manufacturer: troemner llc  72174
 View more versions of this product
Catalog No. 02-216-008
  • / Each
Description & Specifications
Specifications
Packaging Quantity Multiple
Weight (Metric) 1mg to 1kg
Mass 2kg, 1kg, 500g, (2)200g, 100g, 50g, (2)20g, 10g, 5g, (2)2g, 1g
Product Type Large metric analytical weight set
Material Alloy 8 Stainless Steel
Includes Stainless Steel cylindrical weight: 1kg, 500g, 2 x 200g, 100g, 50g, 2 x 20g, 10g, 5g, 2 x 2g, 1g, Stainless Steel sheet weight: 500mg, 2 x 200mg, 100mg, 50mg, 2 x 20mg, 10mg, 5mg, Aluminium sheet weight: 2 x 2mg, 1mg, Polycarbonate case in Wood case
Certifications/Compliance Statement of accuracy, ISO9001
Classification ASTM Class 4
Quantity 25/Set