Please login to your online account to display your discounted pricing
Troemner™ Alloy 8 Metric Precision Weight Sets, Class 4 with NVLAP Certificate
Excellent for high-precision calibrations and reference standards
Manufacturer: troemner llc  72144W
 View more versions of this product
Catalog No. 02-216-015
  • / Each
Description & Specifications
Specifications
Packaging Quantity Multiple
Classification ASTM Class 4
Material Stainless Steel
Includes Cylindrical weight: 2kg, 1kg, 500g, 2 x 200g, 100g, 50g, 2 x 20g, 10g, 5g, 2 x 2g, 1g, Polycarbonate case in Wood case
Product Type Large metric analytical weight set
Quantity 14/Set
Weight (Metric) 1g to 2kg
Certifications/Compliance NVLAP, ISO9001
Mass 2kg, 1kg, 500g, (2)200g, 100g, 50g, (2)20g, 10g, 5g, (2)2g, 1g