missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Troemner™ Alloy 8 Metric Precision Weight Sets, Class 3
Excellent for high-precision calibrations and reference standards
$3,026.34 - $3,249.28
Specifications
Product Type | Medium Metric Analytical Weight Set |
---|---|
Classification | ASTM Class 3 |
Packaging Quantity | Multiple |
Catalog Number | Mfr. No. | Material | Quantity | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Material | Quantity | Price | Quantity | ||||
02-215-20C
|
Troemner™
80859319 |
Stainless Steel, Aluminum | 21/Set |
Each for $3,249.28
|
|
02-215-20B
|
Troemner™
80859289 |
Stainless Steel | 17/Set |
Each for $3,026.34
|
|
Description
Manufactured from a specially developed austenitic stainless-steel—Alloy 8—available exclusively from Troemner. Weights 1g and larger are manufactured using Alloy 8, which possesses closely controlled density, extremely low magnetic susceptibility, excellent stability and resistance to corrosion, assuring a very stable mass over time. Other weights are stainless-steel.
- Each fractional weight is embossed with its value
- Weights from 1 to 30mg are aluminum and flat, with one side turned up to form a handle for non-slip gripping with forceps
- Weights from 50 to 500mg are stainless steel and flat with one side turned up to form a handle
- Gram and kilogram weights are manufactured using Alloy 8 and come in cylindrical shapes with knobs on top for easy handling
- Weight sets come in a 5-2-2-1 series, with two weights of any included mg or g value beginning with “2” (2mg, 20mg, etc.) and one weight for all other inclusive values
Specifications
Medium Metric Analytical Weight Set | |
ASTM Class 3 | |
Multiple |