Troemner™ Alloy 8 Metric Precision Weight Sets, Class 2
Excellent for high-precision calibrations and reference standards
Manufacturer:  Troemner™ 72312DUP
 View more versions of this product
Catalog No. 02-214-056
Description
Description
Designed for your most demanding weighing applications. Analytical Precision Weights meet or exceed specifications as defined in the ANSI/ASTM E617 standard.
  • Low magnetic susceptibility, uniform consistency and high resistance to corrosion
  • Used in extreme stable environments
  • 1 to 500mg weights are one-piece construction with no adjusting cavity with one end turned up for easy handling
  • Weight sets 1mg-500g are provided in a high quality polycarbonate set case
  • Weight sets where the largest weight is 1kg and above are supplied in a felt line oak case
  • Each polycarbonate case is placed in its own individual compartment
Certifications
A NIST/NVLAP™ weight calibration certificate or traceable certificate may be purchased to meet all of your ISO™, GLP, GMP, FDA, or nuclear requirements
1Set includes two 200g weights. 2Set includes two 2kg weights.
Specifications
Specifications
Medium Metric Analytical Weight Set
ASTM Class 2
10 mg to 50 g
Cylindrical weight: 50g, 2 x 20g, 10g, 5g, 2 x 2g, 1g, sheet weight: 500mg, 2 x 200mg, 100mg, 50mg, 2 x 20mg, 10mg, Polycarbonate case
Multiple
Statement of accuracy, ISO9001
Stainless Steel
10 mg to 50 g
16/Set