Please login to your online account to display your discounted pricing

Troemner™ Alloy 8 Metric Precision Weight Sets, Class 2 with Statement of Accuracy


Excellent for high-precision calibrations and reference standards

Manufacturer: fisher scientific  72292F

 View more versions of this product

Catalog No. 02-217-1A

  • / Each
 

Description & Specifications

Specifications

Product Type Medium metric analytical weight set
Certifications/Compliance Statement of accuracy, ISO9001
Includes Cylindrical weight: 50g, 2 x 20g, 10g, 5g, 2 x 2g, 1g, Polycarbonate case
Classification ASTM Class 2
Quantity 8/Set
Packaging Quantity Multiple
Material Stainless Steel
Weight (Metric) 1g to 50g
Mass 1g to 50g