Please login to your online account to display your discounted pricing
Troemner™ Alloy 8 Metric Precision Weight Sets, Class 2 with Statement of Accuracy
Excellent for high-precision calibrations and reference standards
Manufacturer: fisher scientific  72282F
 View more versions of this product
Catalog No. 02-216
  • / Each
Description & Specifications
Specifications
Includes Stainless Steel cylindrical weight: 100g, 50g, 2 x 20g, 10g, 5g, 2 x 2g, 1g, Stainless Steel sheet weight: 500mg, 2 x 200mg, 100mg, 50mg, 2 x 20mg, 10mg, 5mg, Aluminium sheet weight: 2 x 2mg, 1mg, Polycarbonate case
Packaging Quantity Multiple
Product Type Medium metric analytical weight set
Certifications/Compliance Statement of accuracy, ISO9001
Classification ASTM Class 2
Quantity 21/Set
Material Alloy 8 Stainless Steel, Aluminum
Weight (Metric) 1mg to 100g
Mass 1mg to 100g
Accessories
Accessories for Catalog Number: 02216
Item Mfr. No. Item Description Price Per Unit Quantity