Please login to your online account to display your discounted pricing
Troemner™ Alloy 8 Metric Precision Weight Sets, Class 1 with Statement of Accuracy
Excellent for high-precision calibrations and reference standards
Manufacturer: troemner llc  72171
 View more versions of this product
Catalog No. 02-216-006
  • / Each
Description & Specifications
Specifications
Product Type Large metric analytical weight set
Packaging Quantity Multiple
Certifications/Compliance Statement of accuracy, ISO9001
Includes Stainless Steel cylindrical weight: 1kg, 500g, 2 x 200g, 100g, 50g, 2 x 20g, 10g, 5g, 2 x 2g, 1g, Stainless Steel sheet weight: 500mg, 2 x 200mg, 100mg, 50mg, 2 x 20mg, 10mg, 5mg, Aluminium sheet weight: 2 x 2mg, 1mg, Polycarbonate case in Wood case
Classification ASTM Class 1
Quantity 25/Set
Material Aluminum
Weight (Metric) 1mg to 1kg
Mass 1kg, 500g, (2)200g, 100g, 50g, (2)20g, 10g, 5g, (2)2g, 1g, 500mg, (2)200mg, 100mg, 50mg, (2)20mg, 10mg, 5mg, (2)2mg, 1mg