Please login to your online account to display your discounted pricing

Troemner™ Alloy 8 Metric Precision Individual Weights, UltraClass with Statement of Accuracy


Excellent for high-precision calibrations and reference standards

Manufacturer: troemner llc  70260

 View more versions of this product

Catalog No. 02-214K

  • / Each
 

Description & Specifications

Specifications

Classification UltraClass
Quantity Each
Weight (Metric) 500mg
Includes Polycarbonate case
Packaging Quantity 1
Product Type One-piece metric analytical sheet weight
Density 7.95g/cm3 at 20°C
Certifications/Compliance Statement of accuracy, ISO9001
Shape Flat with one end turned up for easy handling
Material 304 Stainless Steel
Mass 500mg
Accessories

Accessories for Catalog Number: 02214K

Item Mfr. No. Item Description Price Per Unit Quantity  
296352992A
Fisherbrand™ Cotton Inspection Gloves
superior glove works ltd
LL40
Ideal for general inspection work or as liners for unlined liquidproof gloves Dozen of 12 Each for $2.99
Case of 100 Dozen for $267.00