Please login to your online account to display your discounted pricing

Troemner™ Alloy 8 Metric Precision Individual Weights, UltraClass with Statement of Accuracy


Excellent for high-precision calibrations and reference standards

Manufacturer: troemner llc  70240*

 View more versions of this product

Catalog No. 02-214H

  • / Each
 

Description & Specifications

Specifications

Product Type Two piece metric analytical weight
Certifications/Compliance Statement of accuracy, ISO9001
Includes Polycarbonate case
Classification UltraClass
Quantity Each
Packaging Quantity 1
Material Alloy 8 Stainless Steel
Weight (Metric) 2g
Density 8.03g/cm3 at 20°C
Mass 2g
Shape Cylindrical with handling knob
Accessories

Accessories for Catalog Number: 02214H

Item Mfr. No. Item Description Price Per Unit Quantity