Please login to your online account to display your discounted pricing

Troemner™ Alloy 8 Metric Precision Individual Weights, UltraClass with Statement of Accuracy


Excellent for high-precision calibrations and reference standards

Manufacturer: troemner llc  70210

 View more versions of this product

Catalog No. 02-214E

  • / Each
 

Description & Specifications

Specifications

Includes Polycarbonate case
Classification UltraClass
Product Type Two piece metric analytical weight
Certifications/Compliance Statement of accuracy, ISO9001
Quantity Each
Packaging Quantity 1
Material Alloy 8 Stainless Steel
Weight (Metric) 10g
Density 8.03g/cm3 at 20°C
Mass 10g
Shape Cylindrical with handling knob
Accessories

Accessories for Catalog Number: 02214E

Item Mfr. No. Item Description Price Per Unit Quantity