missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Troemner™ Alloy 8 Metric Precision Individual Weights, UltraClass

Excellent for high-precision calibrations and reference standards

Manufacturer:  Troemner™ 70140*

 View more versions of this product

Catalog No. 02-215-94



Includes: Forceps

Description

Description

Certifications

A NIST/NVLAP™ weight calibration certificate or traceable certificate may be purchased to meet all of your ISO™, GLP, GMP, FDA, or nuclear requirements

Specifications

Specifications

Two-Piece Metric Analytical Weight
UltraClass
500g
500g
Polycarbonate case
1
Statement of accuracy, ISO9001
Stainless Steel
8.03g/cm3 at 20°C
Cylindrical with handling knob
Each
Documents
Accessories