Please login to your online account to display your discounted pricing

Troemner™ Alloy 8 Metric Precision Individual Weights, UltraClass with Statement of Accuracy


Excellent for high-precision calibrations and reference standards

Manufacturer: troemner llc  70370*

 View more versions of this product

Catalog No. 02-214X

  • / Each
 

Description & Specifications

Specifications

Classification UltraClass
Packaging Quantity 1
Material 3003-H14 Aluminum
Includes Polycarbonate case
Mass 1mg
Shape Flat with one end turned up for easy handling
Quantity Each
Certifications/Compliance Statement of accuracy, ISO9001
Weight (Metric) 1mg
Density 2.7g/cm3 at 20°C
Product Type One-piece metric analytical sheet weight
Accessories

Accessories for Catalog Number: 02214X

Item Mfr. No. Item Description Price Per Unit Quantity  
296352992A
Fisherbrand™ Cotton Inspection Gloves
superior glove works ltd
LL40
Ideal for general inspection work or as liners for unlined liquidproof gloves Dozen of 12 Each for $2.99
Case of 100 Dozen for $267.00