Troemner™ Alloy 8 Metric Precision Individual Weights, UltraClass with Statement of Accuracy
Excellent for high-precision calibrations and reference standards
Manufacturer:  Troemner Llc 70300F
 View more versions of this product
Catalog No. 02-214P
Specifications Description & Specifications
Specifications
One-piece metric analytical sheet weight
UltraClass
50mg
50mg
Polycarbonate case
1
Statement of accuracy, ISO9001
Stainless Steel
7.95g/cm3 at 20°C
Flat with one end turned up for easy handling
Each
Accessories