Please login to your online account to display your discounted pricing

Alfa Aesar™ Trivinylmethylsilane, tech. 90%

Chemical Identifiers

Chemical Name or Material Trivinylmethylsilane
CAS 18244-95-6
Purity Grade Technical Grade
Molecular Formula C7H12Si
Formula Weight 124.26
MDL Number MFCD00051543
UN Number UN1993
Beilstein 1739031
Synonym Methyltrivinylsilane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0607203

  SDS
alfa aesar™
L0607203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0607206

  SDS
alfa aesar™
L0607206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 124.26
Molecular Formula C7H12Si
Health Hazard 3 GHS P Statement
P210-P261-P243
Keep away from heat/sparks/open flames/hot surfaces. - No Smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Take precautionary measures against static discharge.
Chemical Name or Material Trivinylmethylsilane
TSCA TSCA
Assay 90%
MDL Number MFCD00051543
CAS 18244-95-6
UN Number UN1993
Synonym Methyltrivinylsilane
Boiling Point 101° to 102°C
Flash Point 15°C (59°F)
Health Hazard 1 Danger
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Purity Grade Technical Grade
Beilstein 1739031
Quantity 1g
Health Hazard 2 GHS H Statement
H225
Highly flammable liquid and vapor.
Safety and Handling

Danger

GHS P Statement
P210-P261-P243
Keep away from heat/sparks/open flames/hot surfaces. - No Smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Take precautionary measures against static discharge.

GHS H Statement
H225
Highly flammable liquid and vapor.