missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Trityl chloride, polymer-supported, 1% cross-linked, 100-200 mesh, 0.5-2.5 mmol/g on poly(styrene-divinylbenzene)

Manufacturer:  Alfa Aesar™ 4439703

 View more versions of this product

Catalog No. AA4439703

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Trityl chloride
Moisture sensitive
1g
76-83-5
MFCD00284810
benzene, 1,1',1-chloromethylidyne tris
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)Cl
278.779
Beads
100 to 200 Mesh
White
Polymer-supported, 1% cross-linked, 0.5 to 2.5mmol/g on poly(styrene-divinylbenzene)
C19H15Cl
UN3261
JBWKIWSBJXDJDT-UHFFFAOYSA-N
[chloro(diphenyl)methyl]benzene
6456

Chemical Identifiers

76-83-5
278.779
JBWKIWSBJXDJDT-UHFFFAOYSA-N
6456
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)Cl
C19H15Cl
MFCD00284810
benzene, 1,1',1-chloromethylidyne tris
[chloro(diphenyl)methyl]benzene
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

Documents