Please login to your online account to display your discounted pricing

Triton™ X-405, MP Biomedicals™


Quantity 500mL

Manufacturer: mp biomedicals  15241190

 View more versions of this product

Catalog No. ICN15241190

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 1966
Physical Form Liquid
Color Clear colorless
Quantity 500mL
Density 1.1g/mL
Physical State Liquid
Molecular Formula C94H182O41
CAS 9002-93-1
Synonym Alkylphenol hydroxypolyethylene
Chemical Name or Material Triton™ X-405
Recommended Storage Store at room temperature

Specifications

Chemical Name or Material Triton™ X-405
CAS 9002-93-1
Physical Form Liquid
Molecular Formula C94H182O41
Formula Weight 1966
Synonym Alkylphenol hydroxypolyethylene
Accessories

Store at room temperature