missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Triton™ X-405, MP Biomedicals™

$52.96 - $63.82

Chemical Identifiers

CAS 9002-93-1
Physical Form Liquid
Molecular Formula C94H182O41
Formula Weight 1966
Synonym Alkylphenol hydroxypolyethylene
Chemical Name or Material Triton™ X-405
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN15241180 Mp Biomedicals
15241180
100mL Each for $52.96
ICN15241190 Mp Biomedicals
15241190
500mL Each for $63.82
Specifications Description & Specifications

Chemical Identifiers

9002-93-1
C94H182O41
Alkylphenol hydroxypolyethylene
Liquid
1966
Triton™ X-405

Specifications

9002-93-1
Liquid
1966
Liquid
100mL
1.1g/mL
C94H182O41
Alkylphenol hydroxypolyethylene
Clear colorless
Triton™ X-405

Safety and Handling

Recommended Storage : Store at room temperature