Please login to your online account to display your discounted pricing

Triton X-405, 70% solution in water, ACROS Organics™

$89.00 - $89.00

Chemical Identifiers

Chemical Name or Material Triton™ X-405
Name Note 70% Solution in Water
CAS 9002-93-1
CAS 7732-18-5
Assay ≥65%
Molecular Formula C94H182O41
Formula Weight 1968.47
MDL Number MFCD01779734
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC215690025

  SDS
thermo fisher scientific
AC215690025
Glass bottle 2.5L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC215692500

  SDS
fisher scientific acros
AC215692500
Glass bottle 250mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC215690010

  SDS
fisher scientific acros
215690010
Glass bottle 1L Each for $89.00
Description & Specifications

Specifications

Physical Form Liquid
Assay ≥65%
Formula Weight 1968.47
Quantity 2.5L
Color Clear pale yellow
Merck Index 15, 6850
Density 1.08g/mL
Chemical Name or Material Triton™ X-405
Molecular Formula C94H182O41
CAS 9002-93-1
Packaging Glass bottle
Name Note 70% Solution in Water
MDL Number MFCD01779734
Health Hazard 2 GHS H Statement:
Harmful to aquatic life with long lasting effects.
CAS 7732-18-5
Health Hazard 3 GHS P Statement:
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful to aquatic life with long lasting effects.