Please login to your online account to display your discounted pricing

Triton™ X-100 Wetting Agent, Ricca Chemical


Quantity 1L
Packaging Natural poly

Manufacturer: ricca chemical company  RIR86985001A

 View more versions of this product

Catalog No. R86985001A

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Triton™ X-100 Wetting Agent
Shelf Life 24 months
CAS 9002-93-1
CAS Min % 0.98
CAS Max % 1.0
Physical Form Viscous Liquid
Packaging Natural poly
Color Colorless
Quantity 1L

Specifications

Chemical Name or Material Triton™ X-100 Wetting Agent
CAS 9002-93-1
CAS Min % 0.98
CAS Max % 1.0
Physical Form Viscous Liquid
Accessories

24 months