missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Triton X-100, Hydrogenated, Protein Grade™, 10% Solution, Sterile-Filter, Calbiochem™,

Supplier:  MilliporeSigma™ 64846410ML

Catalog No. 64846410ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Colorless
10 mL
Octylphenoxypolyethoxyethanol, Hydrogenated
101013-07-4
Plastic Ampoule
Protein

Chemical Identifiers

101013-07-4
Octylphenoxypolyethoxyethanol, Hydrogenated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions