Please login to your online account to display your discounted pricing

Triton X-100, EMD-Millipore


Quantity 1L
Packaging Glass

Manufacturer: emd millipore corporation  1122981001

 View more versions of this product

Catalog No. M1122981001

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 1L
Density Density: 1.07g/cm3 (20°)
Chemical Name or Material Triton X-100
Synonym Octoxynol, (p-tert-Octylphenoxy)polyethoxyethanol, O-[4-(1,1,3,3-Tetramethylbutyl)phenoxy]polyethoxyethanol, Polyethyleneglycol-[4-(1,1,3,3-tetramethylbutyl)phenyl]-ether
Linear Formula C8H17C6H4(OCH2CH2)nOH
Recommended Storage Store without restrictions
Molecular Formula C14H21(C2H4O)nOH
Packaging Glass
CAS 9036-19-5
Vapor Pressure < - 0.01 hPa (20°C)

Specifications

Chemical Name or Material Triton X-100
CAS 9036-19-5
Molecular Formula C14H21(C2H4O)nOH
Linear Formula C8H17C6H4(OCH2CH2)nOH
Synonym Octoxynol, (p-tert-Octylphenoxy)polyethoxyethanol, O-[4-(1,1,3,3-Tetramethylbutyl)phenoxy]polyethoxyethanol, Polyethyleneglycol-[4-(1,1,3,3-tetramethylbutyl)phenyl]-ether
Accessories

Store without restrictions