missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Triton X-100, EMD-Millipore
$254.41 - $1,183.79
Chemical Identifiers
CAS | 9036-19-5 |
---|---|
Molecular Formula | C14H21(C2H4O)nOH |
Synonym | Octoxynol, (p-tert-Octylphenoxy)polyethoxyethanol, O-[4-(1,1,3,3-Tetramethylbutyl)phenoxy]polyethoxyethanol, Polyethyleneglycol-[4-(1,1,3,3-tetramethylbutyl)phenyl]-ether |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
M1122980101
|
Emd Millipore Corporation
1122980101 |
100mL | Glass |
Each for $254.41
|
|
M1122981001
|
Emd Millipore Corporation
1122981001 |
1L | Glass |
Each for $1,183.79
|
|
Chemical Identifiers
9036-19-5 | |
C14H21(C2H4O)nOH | |
Octoxynol, (p-tert-Octylphenoxy)polyethoxyethanol, O-[4-(1,1,3,3-Tetramethylbutyl)phenoxy]polyethoxyethanol, Polyethyleneglycol-[4-(1,1,3,3-tetramethylbutyl)phenyl]-ether |
Specifications
9036-19-5 | |
C8H17C6H4(OCH2CH2)nOH | |
Glass | |
Density: 1.07g/cm3 (20°) | |
Triton X-100 |
C14H21(C2H4O)nOH | |
Octoxynol, (p-tert-Octylphenoxy)polyethoxyethanol, O-[4-(1,1,3,3-Tetramethylbutyl)phenoxy]polyethoxyethanol, Polyethyleneglycol-[4-(1,1,3,3-tetramethylbutyl)phenyl]-ether | |
< - 0.01 hPa (20°C) | |
100mL |
Safety and Handling
Recommended Storage : Store without restrictions