Please login to your online account to display your discounted pricing

Triton™ X-100, ACROS Organics™


Quantity 1L
Packaging EcoSafPak™

Manufacturer: thermo fisher scientific  215680010

 View more versions of this product

Catalog No. AC215680010

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 646.85
Quantity 1L
Melting Point 6.0°C
UN Number 3082
Flash Point 274°C
Boiling Point 270.0°C
Merck Index 15, 6850
Packaging EcoSafPak™
Density 1.0670g/mL
Synonym Polyoxyethylene(10) octylphenyl ether
CAS 9002-93-1
Health Hazard 1 Danger
Chemical Name or Material Triton™ X-100
Molecular Formula C34H62O11
MDL Number MFCD00132505
Health Hazard 3 Emergency Overview
Harmful if swallowed. Risk of serious damage to eyes. Harmful to aquatic organisms, may cause long-term adverse effects in the aquatic environment.
NFPA&C
Health Hazard 2 GHS H Statement:
Harmful to aquatic life with long lasting effects.
Causes serious eye damage.
Harmful if swallowed.

Specifications

Chemical Name or Material Triton™ X-100
CAS 9002-93-1
Molecular Formula C34H62O11
Formula Weight 646.85
MDL Number MFCD00132505
Synonym Polyoxyethylene(10) octylphenyl ether
Accessories

Danger

Emergency Overview
Harmful if swallowed. Risk of serious damage to eyes. Harmful to aquatic organisms, may cause long-term adverse effects in the aquatic environment.
NFPA&C

GHS H Statement:
Harmful to aquatic life with long lasting effects.
Causes serious eye damage.
Harmful if swallowed.

SDS & Safety Documents