missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Triton™ X-100, ACROS Organics™

$45.91 - $757.65

Chemical Identifiers

CAS 9002-93-1
Molecular Formula C16H26O2
Molecular Weight (g/mol) 250.382
MDL Number MFCD00132505
InChI Key JYCQQPHGFMYQCF-UHFFFAOYSA-N
Synonym Polyoxyethylene(10) octylphenyl ether
PubChem CID 5590
IUPAC Name 2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethanol
SMILES CC(C)(C)CC(C)(C)C1=CC=C(C=C1)OCCO
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC215682500
SDS View Documents
Thermo Fisher Scientific
215682500
250mL Glass Bottle
Each for $45.91
 
AC215680010
SDS View Documents
Thermo Fisher Scientific
215680010
1L EcoSafPak™
Each for $105.89
 
AC215680100
SDS View Documents
Thermo Fisher Scientific
AC215680100
10L Plastic drum
Each for $757.65
 
Description

Description

Specifications

Chemical Identifiers

9002-93-1
250.382
JYCQQPHGFMYQCF-UHFFFAOYSA-N
5590
CC(C)(C)CC(C)(C)C1=CC=C(C=C1)OCCO
C16H26O2
MFCD00132505
Polyoxyethylene(10) octylphenyl ether
2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethanol

Specifications

9002-93-1
C16H26O2
3082
Polyoxyethylene(10) octylphenyl ether
Solubility in water: soluble. Other solubilities: soluble in toluene, xylene, trichloroethylene,, ethylene glycol, ethyl ether, ethyl alcohol,, isopropyl alcohol, ethylene dichloride and most, other solvnets., insoluble in kerosene, miniral spirits.
CC(C)(C)CC(C)(C)C1=CC=C(C=C1)OCCO
250.382
1.4900 to 1.4920
240 mPa.s (25°C)
6.0°C
250mL
Triton™ X-100
1.0670g/mL
MFCD00132505
15, 6850
Glass Bottle
JYCQQPHGFMYQCF-UHFFFAOYSA-N
2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethanol
5590
646.85
270.0°C
274°C
1.067
Safety and Handling

Safety and Handling

GHS H Statement
Harmful to aquatic life with long lasting effects.
Causes serious eye damage.
Harmful if swallowed.

GHS P Statement
Avoid release to the environment.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

Danger

SDS