missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Trisodium N-(1-Carboxylatoethyl)iminodiacetate Hydrate, TCI America™

Manufacturer:  TCI America T220225G

 View more versions of this product

Catalog No. T220225G



Specifications

Specifications

Trisodium N-(1-Carboxylatoethyl)iminodiacetate Hydrate
25g
C7H11NNa3O6
SQKOOOSPGWXVRN-UHFFFAOYSA-N
2-[bis(carboxymethyl)amino]propanoic acid;sodium
131849341
Crystal-Powder at 20°C
White-Yellow
164462-16-2
n,n-bis carboxylatomethyl alanine trisodium salt
CC(C(=O)O)N(CC(=O)O)CC(=O)O.[Na].[Na].[Na]
274.135
271.11
≥95.0% (T)

Chemical Identifiers

164462-16-2
274.135
n,n-bis carboxylatomethyl alanine trisodium salt
2-[bis(carboxymethyl)amino]propanoic acid;sodium
C7H11NNa3O6
SQKOOOSPGWXVRN-UHFFFAOYSA-N
131849341
CC(C(=O)O)N(CC(=O)O)CC(=O)O.[Na].[Na].[Na]
Safety and Handling

Safety and Handling

TSCA : Yes

Documents