missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Trisodium Hydrogen Ethylenediaminetetraacetate Hydrate, TCI America™

Manufacturer:  TCI America E0430500G

 View more versions of this product

Catalog No. E0430500G



Specifications

Specifications

Trisodium Hydrogen Ethylenediaminetetraacetate Hydrate
500g
C10H15N2Na3O9
trisodium hydrogen ethylenediaminetetraacetate hydrate, tricon bw, unii-zmn5a047nl, endrate, edta triasodium salt, zmn5a047nl, ethylenediaminetetraacetic acid trisodium salt hydrate, sequestrene sodium 3, trisodium edta monohydrate, edetate trisodium monohydrate
C(CN(CC(=O)[O-])CC(=O)[O-])N(CC(=O)O)CC(=O)[O-].O.[Na+].[Na+].[Na+]
376.204
358.19
≥98.0% (T)
White
85715-60-2
MFCD00013305
KKTNQJMZBPLVKM-UHFFFAOYSA-K
trisodium;2-[2-[bis(carboxylatomethyl)amino]ethyl-(carboxymethyl)amino]acetate;hydrate
16211179
Crystal-Powder at 20°C

Chemical Identifiers

85715-60-2
376.204
KKTNQJMZBPLVKM-UHFFFAOYSA-K
16211179
C(CN(CC(=O)[O-])CC(=O)[O-])N(CC(=O)O)CC(=O)[O-].O.[Na+].[Na+].[Na+]
C10H15N2Na3O9
MFCD00013305
trisodium hydrogen ethylenediaminetetraacetate hydrate, tricon bw, unii-zmn5a047nl, endrate, edta triasodium salt, zmn5a047nl, ethylenediaminetetraacetic acid trisodium salt hydrate, sequestrene sodium 3, trisodium edta monohydrate, edetate trisodium monohydrate
trisodium;2-[2-[bis(carboxylatomethyl)amino]ethyl-(carboxymethyl)amino]acetate;hydrate
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1265

RTECSNumber : AH5250000

TSCA : No

Documents