missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Trisodium citrate, anhydrous, 99%

Manufacturer:  Alfa Aesar™ 4555630

 View more versions of this product

Catalog No. AA4555630



Specifications

Specifications

Trisodium citrate
White
250g
68-04-2
MFCD00012462
sodium citrate, trisodium citrate, citrosodine, sodium citrate anhydrous, citrosodina, natrocitral, citnatin, citreme, citrosodna, trisodium citrate, anhydrous
C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.[Na+].[Na+].[Na+]
258.068
CHEBI:53258
Solid
Hygroscopic
300°C
Anhydrous
C6H5Na3O7
14,8602
HRXKRNGNAMMEHJ-UHFFFAOYSA-K
trisodium;2-hydroxypropane-1,2,3-tricarboxylate
6224
258.07
99%

Chemical Identifiers

68-04-2
258.068
HRXKRNGNAMMEHJ-UHFFFAOYSA-K
6224
trisodium;2-hydroxypropane-1,2,3-tricarboxylate
C6H5Na3O7
MFCD00012462
sodium citrate, trisodium citrate, citrosodine, sodium citrate anhydrous, citrosodina, natrocitral, citnatin, citreme, citrosodna, trisodium citrate, anhydrous
CHEBI:53258
C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.[Na+].[Na+].[Na+]
Safety and Handling

Safety and Handling

RTECSNumber : GE8300000

TSCA : TSCA

SDS Documents