Please login to your online account to display your discounted pricing

Alfa Aesar™ Trisodium citrate, anhydrous, 99%

$33.30 - $98.24

Chemical Identifiers

Chemical Name or Material Trisodium citrate
CAS 68-04-2
Name Note Anhydrous
Formula Weight 258.07
MDL Number MFCD00012462
Merck Index 14,8602
Synonym Citric acid trisodium salt, anhydrous
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4555630

  SDS
alfa aesar™
4555630
250g Each for $33.30

AA45556A1

  SDS
alfa aesar™
45556A1
1kg Each for $98.24

AA45556A7

  SDS
alfa aesar™
45556A7
5kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 258.07
Name Note Anhydrous
MDL Number MFCD00012462
Merck Index 14,8602
CAS 68-04-2
Quantity 250g
Synonym Citric acid trisodium salt, anhydrous
RTECS Number GE8300000
Chemical Name or Material Trisodium citrate
Melting Point 300°C
Physical Form Solid
Assay 99%
TSCA TSCA
Color White
Sensitivity Hygroscopic