missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

TrisHydroxymethyl)Aminomethane ACS AR, Macron Fine Chemicals™

macron

Chemical Identifiers

CAS 77-86-1
Molecular Formula NH2C(CH2OH)3
Formula Weight 121.14
Chemical Name or Material Tris(Hydroxymethyl)Aminomethane
Grade ACS Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
MK1806100 Avantor Performance
180657
Glass 100g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
MK18065 Avantor Performance
180611
STAKMOR 5kg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
MK1806500 Avantor Performance
180610*
Glass 500g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

77-86-1
121.14
ACS Grade
NH2C(CH2OH)3
Tris(Hydroxymethyl)Aminomethane

Specifications

121.14
Glass
100g
ACS Grade
77-86-1
NH2C(CH2OH)3
Tris(Hydroxymethyl)Aminomethane