missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tris (Tromethamine) (Granular/USP), Fisher Chemical

Manufacturer:  Thermo Fisher Scientific FLT39312

 View more versions of this product

Catalog No. T393-12


 

Specifications Description & Specifications

Specifications

121.14
219°C
12kg
USP
C4H11NO3
1.0% max. (USP)
0.001% max. (USP)
Meets Requirements (USP)
MFCD00004679
Pass Test (USP)
H2NC(CH2OH)3
Tris (Tromethamine)
168.5°C
Powder/Solid
99.0 to 101.0% (USP)
White
10.4
0.1% max. (USP)
THAM™
Poly Pail
168° to 172°C (USP)
168° to 172°C (USP)
77-86-1

Chemical Identifiers

77-86-1
C4H11NO3
121.14
THAM™
99.0 to 101.0% (USP)
Powder/Solid
H2NC(CH2OH)3
MFCD00004679
Tris (Tromethamine)
USP

Safety and Handling

SDS & Safety Documents