Please login to your online account to display your discounted pricing

Tris(triphenylphosphine)ruthenium(II) chloride, 98%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC207880250

 View more versions of this product

Catalog No. AC207880250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Tris(triphenylphosphine)ruthenium(II) chloride
CAS 15529-49-4
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Carbon (C) 66.0 to 69.0 %
Infrared Spectrum Authentic
Physical Form Solution
Assay ≥10.3% (Ru)
Assay Percent Range 98%
Molecular Formula C54H45Cl2P3Ru
Linear Formula {(C6H5 )3P}3RuCl2
Packaging Glass bottle
Formula Weight 958.85
MDL Number MFCD00013077
Synonym Dichlorotris(triphenylphosphine)ruthenium(II)
Color Clear
Quantity 25g

Specifications

Chemical Name or Material Tris(triphenylphosphine)ruthenium(II) chloride
Assay ≥10.3% (Ru)
Assay Percent Range 98%
Molecular Formula C54H45Cl2P3Ru
Linear Formula {(C6H5 )3P}3RuCl2
Formula Weight 958.85
MDL Number MFCD00013077
Synonym Dichlorotris(triphenylphosphine)ruthenium(II)
Accessories

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.

SDS & Safety Documents