Please login to your online account to display your discounted pricing

Tris(triphenylphosphine)rhodium(I) chloride, 99.99%, (trace metal basis), ACROS Organics™


Quantity 250mg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC194572500

 View more versions of this product

Catalog No. AC194572500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Purity Grade Trace Metal Basis
Formula Weight 925.23
Quantity 250mg
Color Maroon
TSCA TSCA
EINECS Number 238-744-5
Merck Index 15, 10245
Fieser 01,140; 02,448; 03,325; 04,559; 05,736; 06,652; 07,68; 08,109; 09,113; 15,90
Packaging Glass bottle
Total Trace Metal Impurities 0.01% max.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Synonym Tris(triphenylphosphine)chlororhodium
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Linear Formula [(C6H5)3P]3RhCl
CAS 14694-95-2
Name Note 99.99%
Molecular Formula C54H45ClP3Rh
Assay Percent Range 99.99%
Chemical Name or Material Tris(triphenylphosphine)rhodium(I) chloride
MDL Number MFCD00010016

Specifications

Chemical Name or Material Tris(triphenylphosphine)rhodium(I) chloride
Name Note 99.99%
CAS 14694-95-2
Purity Grade Trace Metal Basis
Assay Percent Range 99.99%
Molecular Formula C54H45ClP3Rh
Linear Formula [(C6H5)3P]3RhCl
Formula Weight 925.23
MDL Number MFCD00010016
Synonym Tris(triphenylphosphine)chlororhodium
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

SDS & Safety Documents