Please login to your online account to display your discounted pricing

Tris(triphenylphosphine)rhodium(I) chloride, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  190420050

 View more versions of this product

Catalog No. AC190420050

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Formula Weight 925.23
Quantity 5g
Color Red-brown to maroon
EINECS Number 238-744-5
TSCA TSCA
Fieser 01,140; 02,448; 03,325; 04,559; 05,736; 06,652; 07,68; 08,109; 09,113; 15,90
Merck Index 15, 10245
Gravimetry 10.8 to 11.3% (Rh)
Infrared Spectrum Authentic
Packaging Glass bottle
Synonym Tris(triphenylphosphine)chlororhodium
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Chemical Name or Material Tris(triphenylphosphine)rhodium(I) chloride
Name Note 98%
Assay Percent Range 98%
Molecular Formula C54H45ClP3Rh
Linear Formula [(C6H5)3P]3RhCl
MDL Number MFCD00010016
CAS 14694-95-2
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Tris(triphenylphosphine)rhodium(I) chloride
Name Note 98%
CAS 14694-95-2
Assay Percent Range 98%
Molecular Formula C54H45ClP3Rh
Linear Formula [(C6H5)3P]3RhCl
Formula Weight 925.23
MDL Number MFCD00010016
Synonym Tris(triphenylphosphine)chlororhodium
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

SDS & Safety Documents