Please login to your online account to display your discounted pricing

Tris(trimethylsilyloxy)silane, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Tris(trimethylsilyloxy)silane
CAS 1873-89-8
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C9H28O3Si4
Formula Weight 296.66
MDL Number MFCD00054865
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC313940250

  SDS
thermo fisher scientific
AC313940250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Formula Weight 296.66
Physical Form Liquid
Quantity 25g
Color Clear colorless
Density 0.8520g/mL
Refractive Index 1.3850 to 1.3870 (20°C, 589nm)
Infrared Spectrum Authentic
MDL Number MFCD00054865
CAS 1873-89-8
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Assay Percent Range 97%
Molecular Formula C9H28O3Si4
Chemical Name or Material Tris(trimethylsilyloxy)silane
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Flammable liquid and vapor.

Reduction reagent